Learn More
Biomatik Corporation Human Hornerin (HRNR) ELISA Kit
Supplier:  Biomatik Corporation EKU04836
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 78-5,000pg/mL. Sensitivity: 32pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q86YZ3. Target Synonyms: S100a18. Restriction: For Research Use Only.
Catalog No. 50-228-6076