Learn More
Biomatik Corporation Mouse Ferritin, Light Polypeptide (FTL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04073
Encompass_Preferred
Mouse Ferritin, Light Polypeptide (FTL) ELISA Kit for detecting FTL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29391. Target Synonyms: Ferritin Light Chain; Ferritin Light Polypeptide-Like 3; L Apoferritin; Ferritin L Subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2469