Learn More
Biomatik Corporation Mouse Ferroportin (FPN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04080
Encompass_Preferred
Mouse Ferroportin (FPN) ELISA Kit for detecting FPN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JHI9. Gene ID: 53945. Target Synonyms: SLC40A1; FPN1; HFE4; IREG1; MTP1; SLC11A3; Solute Carrier Family 40,Member 1; Iron-Regulated Transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2471