Learn More
Biomatik Corporation Human Fetal Hemoglobin (HBF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04083
Encompass_Preferred
The Human Fetal Hemoglobin (HBF) ELISA Kit allows for quantitative determination of Fetal Hemoglobin in Serum, Plasma, Erythrocyte lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 2.47-200 ng/mL. Sensitivity: 0.99 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Fetal Hemoglobin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2473