Learn More
Biomatik Corporation Bovine Immunoglobulin M (IgM) ELISA Kit
Supplier:  Biomatik Corporation EKU04909
Encompass_Preferred
Bovine Immunoglobulin M (IgM) ELISA Kit for detecting IgM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.71ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: BC114850.1. Target Synonyms: IGHM; Immunoglobulin Heavy Constant Mu; Ig mu chain C region. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6092