Learn More
Biomatik Corporation Mouse Inhibin Beta C (INHbC) ELISA Kit
Supplier:  Biomatik Corporation EKU04937
Encompass_Preferred
Mouse Inhibin Beta C (INHbC) ELISA Kit for detecting INHbC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20pg/ml. Sensitivity: 0.119pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55104. Gene ID: 16325. Target Synonyms: IHBC; INH-Bc. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6096