Learn More
Biomatik Corporation Human Fibrinogen (FG) ELISA Kit, 96 tests
Catalog No. 501502476
Encompass_Preferred
The Human Fibrinogen (FG) ELISA Kit allows for quantitative determination of Fibrinogen in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 12.50-800 ng/mL. Sensitivity: 5.6 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: ibrino. Cross Activity: No significant cross-reactivity or interference between Fibrinogen and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2476 Supplier Biomatik Corporation Supplier No. EKU04090
The Human Fibrinogen (FG) ELISA Kit allows for quantitative determination of Fibrinogen in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 12.50-800 ng/mL. Sensitivity: 5.6 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: ibrino. Cross Activity: No significant cross-reactivity or interference between Fibrinogen and analogues was observed. Restriction: For Research Use Only.