Learn More
Biomatik Corporation Bovine Insulin Like Growth Factor Binding Protein 2 (IGFBP2) ELISA Kit
Supplier:  Biomatik Corporation EKU04988
Encompass_Preferred
Sample Type: Serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Bovine. Detection Range: 0.94-60ng/mL. Sensitivity: 0.32ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P13384. Target Synonyms: IBP2; IGF-BP53. Restriction: For Research Use Only.
Catalog No. 50-228-6100