Learn More
Biomatik Corporation Rat Fibrinogen Beta (FGb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04098
Encompass_Preferred
Rat Fibrinogen Beta Chain (FGB) ELISA Kit for detecting FGB in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.73ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14480. Gene ID: 24366. Target Synonyms: FG-B; Fibrinopeptide B; Fibrinogen Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2479