Learn More
Biomatik Corporation Human Fibrinogen Like Protein 1 (FGL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04107
Encompass_Preferred
The Human Fibrinogen Like Protein 1 (FGL1) ELISA Kit allows for quantitative determination of Fibrinogen Like Protein 1 in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.074 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q08830. Cross Activity: No significant cross-reactivity or interference between Fibrinogen Like Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2481