Learn More
Biomatik Corporation Mouse Fibrinogen Like Protein 2 (FGL2) ELISA Kit, 96 tests
Catalog No. 501502484
Encompass_Preferred
Mouse Fibrinogen Like Protein 2 (FGL2) ELISA Kit for detecting FGL2 in plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.135ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12804. Gene ID: 14190. Target Synonyms: T49; PT49; Fibroleukin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2484 Supplier Biomatik Corporation Supplier No. EKU04110
Mouse Fibrinogen Like Protein 2 (FGL2) ELISA Kit for detecting FGL2 in plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.135ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12804. Gene ID: 14190. Target Synonyms: T49; PT49; Fibroleukin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.