Learn More
Biomatik Corporation Mouse Fibroblast Activation Protein Alpha (FAPa) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU04118
Mouse Fibroblast Activation Protein Alpha (FAPa) ELISA Kit for detecting FAPa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97321. Gene ID: 14089. Target Synonyms: APCE; SIMP; DPPIV; Seprase; 170 kDa melanoma membrane-bound gelatinase; Post-proline cleaving enzyme; Serine integral membrane protease; Surface-expressed protease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.