Learn More
Biomatik Corporation Bovine Interleukin 4 (IL4) ELISA Kit
Supplier:  Biomatik Corporation EKU05329
Encompass_Preferred
Bovine Interleukin 4 (IL4) ELISA Kit for detecting IL4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30367. Gene ID: 280824. Target Synonyms: BSF1; BCGF1 ;B Cell Stimulatory Factor 1; Lymphocyte stimulatory factor 1; B Cell Growth Factor; Binetrakin; Pitrakinra. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6146