Learn More
Biomatik Corporation Pig Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit, 96 tests
Catalog No. 501502494
Encompass_Preferred
Pig Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit for detecting FGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.09pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q60487. Target Synonyms: B-FGF; BFGF; FGFB; HBGH-2; Basic Fibroblast Growth Factor; Heparin-binding growth factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2494 Supplier Biomatik Corporation Supplier No. EKU04155
Pig Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit for detecting FGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.09pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q60487. Target Synonyms: B-FGF; BFGF; FGFB; HBGH-2; Basic Fibroblast Growth Factor; Heparin-binding growth factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.