Learn More
Biomatik Corporation Human Cytokeratin 14 (CK14) ELISA Kit
Supplier:  Biomatik Corporation EKU05464
Encompass_Preferred
Human Cytokeratin 14 (CK14) ELISA Kit for detecting CK14 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02533. Gene ID: 3861. Target Synonyms: KRT14; EBS3; EBS4; K14; Keratin 14; Epidermolysis Bullosa Simplex Dowling-Meara Koebner; Keratin, type I cytoskeletal 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6164