Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit, 96 tests
Catalog No. 501502495
Encompass_Preferred
Rat Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit for detecting FGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.11pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P13109. Gene ID: 54250. Target Synonyms: B-FGF; BFGF; FGFB; HBGH-2; Basic Fibroblast Growth Factor; Heparin-binding growth factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2495 Supplier Biomatik Corporation Supplier No. EKU04156
Rat Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit for detecting FGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.11pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P13109. Gene ID: 54250. Target Synonyms: B-FGF; BFGF; FGFB; HBGH-2; Basic Fibroblast Growth Factor; Heparin-binding growth factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.