Learn More
Biomatik Corporation Mouse Klotho Beta (KLb) ELISA Kit
Supplier:  Biomatik Corporation EKU05514
Encompass_Preferred
Mouse Klotho Beta (KLb) ELISA Kit for detecting KLb in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99N32. Gene ID: 83379. Target Synonyms: Klotho beta-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6171