Learn More
Biomatik Corporation Pig Fibroblast Growth Factor 21 (FGF21) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04159
Encompass_Preferred
Pig Fibroblast Growth Factor 21 (FGF21) ELISA Kit for detecting FGF21 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: C6JWG4. Gene ID: 100302504. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2497