Learn More
Biomatik Corporation Human Leukemia Inhibitory Factor Receptor (LIFR) ELISA Kit
Supplier:  Biomatik Corporation EKU05617
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.117ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P42702. Target Synonyms: CD118; SJS2; STWS; SWS. Restriction: For Research Use Only.
Catalog No. 50-228-6187