Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 9 (FGF9) ELISA Kit, 96 tests
Catalog No. 501502507
Encompass_Preferred
Mouse Fibroblast Growth Factor 9 (FGF9) ELISA Kit for detecting FGF9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P54130. Gene ID: 14180. Target Synonyms: GAF; HBFG9; Glia-Activating Factor; Heparin-Binding Growth Factor 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2507 Supplier Biomatik Corporation Supplier No. EKU04186
Mouse Fibroblast Growth Factor 9 (FGF9) ELISA Kit for detecting FGF9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P54130. Gene ID: 14180. Target Synonyms: GAF; HBFG9; Glia-Activating Factor; Heparin-Binding Growth Factor 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.