Learn More
Biomatik Corporation Mouse Low Density Lipoprotein (LDL) ELISA Kit
Supplier:  Biomatik Corporation EKU05675
Encompass_Preferred
Mouse Low Density Lipoprotein (LDL) ELISA Kit for detecting LDL in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000ng/ml. Sensitivity: 26.5ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: E9Q414. Gene ID: 238055. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6196