Learn More
Biomatik Corporation Mouse Fibronectin (FN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04200
Encompass_Preferred
Mouse Fibronectin (FN) ELISA Kit for detecting FN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11276. Gene ID: 14268. Target Synonyms: FN1; CIG; FINC; LETS; MSF; GFND2; Anastellin; Migration-Stimulating Factor; Cold-Insoluble Globulin; Large, External, Transformation-Sensitive Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2512