Learn More
Biomatik Corporation Human Fibronectin Type III Domain Containing Protein 5 (FNDC5) ELISA Kit, 96 tests
Catalog No. 501502514
Encompass_Preferred
The Human Fibronectin Type III Domain Containing Protein 5 (FNDC5) ELISA Kit allows for quantitative determination of Fibronectin Type III Domain Containing Protein 5 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8NAU1. Cross Activity: No significant cross-reactivity or interference between Fibronectin Type III Domain Containing Protein 5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2514 Supplier Biomatik Corporation Supplier No. EKU04204
The Human Fibronectin Type III Domain Containing Protein 5 (FNDC5) ELISA Kit allows for quantitative determination of Fibronectin Type III Domain Containing Protein 5 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8NAU1. Cross Activity: No significant cross-reactivity or interference between Fibronectin Type III Domain Containing Protein 5 and analogues was observed. Restriction: For Research Use Only.