Learn More
Biomatik Corporation Human Fibulin 1 (FBLN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04207
Encompass_Preferred
The Human Fibulin 1 (FBLN1) ELISA Kit allows for quantitative determination of Fibulin 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.34 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P23142. Cross Activity: No significant cross-reactivity or interference between Fibulin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2515