Learn More
Biomatik Corporation Human Fibulin 3 (FBLN3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04211
Encompass_Preferred
The Human Fibulin 3 (FBLN3) ELISA Kit allows for quantitative determination of Fibulin 3 in Serum, Plasma, Urine and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.64 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q12805. Cross Activity: No significant cross-reactivity or interference between Fibulin 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2518