Learn More
Biomatik Corporation 25-Hydroxyvitamin D3 (HVD3) ELISA Kit
Supplier:  Biomatik Corporation EKU02021
Encompass_Preferred
25-Hydroxyvitamin D3 (HVD3) ELISA Kit for detecting HVD3 in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 2.47-200ng/ml. Sensitivity: 0.91ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: 25-HVD3; 25HVD3; Calcifediol; Calcidiol; H-VD3; 25-Hydroxycholecalciferol; 25-Hydroxy Vitamin D3; 25 (OH) D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5751