Learn More
Biomatik Corporation Human Acylation Stimulating Protein (ASP) ELISA Kit
Supplier:  Biomatik Corporation EKU02121
Encompass_Preferred
Human Acylation Stimulating Protein (ASP) ELISA Kit for detecting ASP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01024. Gene ID: 718. Target Synonyms: Acyl Stimulation Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5761