Learn More
Biomatik Corporation Human Ficolin 1 (FCN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04216
Encompass_Preferred
The Human Ficolin 1 (FCN1) ELISA Kit allows for quantitative determination of Ficolin 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.066 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O00602. Cross Activity: No significant cross-reactivity or interference between Ficolin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2521