Learn More
Biomatik Corporation Dog Adrenocorticotropic Hormone (ACTH) ELISA Kit
Supplier:  Biomatik Corporation EKU02175
Encompass_Preferred
Dog Adrenocorticotropic Hormone (ACTH) ELISA Kit for detecting ACTH in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 12.35-1000pg/ml. Sensitivity: 5.06pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: E2RQ39. Target Synonyms: Corticotropin; Adrenocorticotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5767