Learn More
Biomatik Corporation Human Ficolin 3 (FCN3) ELISA Kit, 96 tests
Catalog No. 501502522
Encompass_Preferred
The Human Ficolin 3 (FCN3) ELISA Kit allows for quantitative determination of Ficolin 3 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 37 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75636. Cross Activity: No significant cross-reactivity or interference between Ficolin 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2522 Supplier Biomatik Corporation Supplier No. EKU04220
The Human Ficolin 3 (FCN3) ELISA Kit allows for quantitative determination of Ficolin 3 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 37 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75636. Cross Activity: No significant cross-reactivity or interference between Ficolin 3 and analogues was observed. Restriction: For Research Use Only.