Learn More
Biomatik Corporation Human Filamin A Alpha (FLNa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04223
Encompass_Preferred
The Human Filamin A Alpha (FLNa) ELISA Kit allows for quantitative determination of Filamin A Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.88-120 ng/mL. Sensitivity: 0.65 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P21333. Cross Activity: No significant cross-reactivity or interference between Filamin A Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2525