Learn More
Biomatik Corporation Mouse Bcl2 Associated X Protein (Bax) ELISA Kit
Supplier:  Biomatik Corporation EKU02680
Encompass_Preferred
Mouse Bcl2 Associated X Protein (Bax) ELISA Kit for detecting Bax in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 25.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07813. Gene ID: 12028. Target Synonyms: BCL2L4; Bax Zeta; Apoptosis regulator BAX; Bcl-2-like protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5831