Learn More
Biomatik Corporation Human FMS Like Tyrosine Kinase 3 (Flt3) ELISA Kit, 96 tests
Catalog No. 501502529
Encompass_Preferred
The Human FMS Like Tyrosine Kinase 3 (Flt3) ELISA Kit allows for quantitative determination of FMS Like Tyrosine Kinase 3 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P36888. Cross Activity: No significant cross-reactivity or interference between FMS Like Tyrosine Kinase 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2529 Supplier Biomatik Corporation Supplier No. EKU04239
The Human FMS Like Tyrosine Kinase 3 (Flt3) ELISA Kit allows for quantitative determination of FMS Like Tyrosine Kinase 3 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P36888. Cross Activity: No significant cross-reactivity or interference between FMS Like Tyrosine Kinase 3 and analogues was observed. Restriction: For Research Use Only.