Learn More
Biomatik Corporation Human Cell Adhesion Molecule 1 (CADM1) ELISA Kit
Supplier:  Biomatik Corporation EKU03062
Encompass_Preferred
Human Cell Adhesion Molecule 1 (CADM1) ELISA Kit for detecting CADM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BY67. Gene ID: 23705. Target Synonyms: BL2; IGSF4; IGSF4A; NECL2; RA175; ST17; SYNCAM; TSLC1; sTSLC-1; synCAM1; Tumor Suppressor In Lung Cancer 1; Immunoglobulin Superfamily,Member 4; Nectin-Like 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5874