Learn More
Biomatik Corporation Human Follicle Stimulating Hormone (FSH) ELISA Kit, 96 tests
Catalog No. 501502531
Encompass_Preferred
The Human Follicle Stimulating Hormone (FSH) ELISA Kit allows for quantitative determination of Follicle Stimulating Hormone in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 mIU/mL. Sensitivity: 0.34 mIU/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01215. Cross Activity: No significant cross-reactivity or interference between Follicle Stimulating Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2531 Supplier Biomatik Corporation Supplier No. EKU04246
The Human Follicle Stimulating Hormone (FSH) ELISA Kit allows for quantitative determination of Follicle Stimulating Hormone in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 mIU/mL. Sensitivity: 0.34 mIU/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01215. Cross Activity: No significant cross-reactivity or interference between Follicle Stimulating Hormone and analogues was observed. Restriction: For Research Use Only.