Learn More
Biomatik Corporation Mouse Complement Factor D (CFD) ELISA Kit
Supplier:  Biomatik Corporation EKU03421
Encompass_Preferred
Mouse Complement Factor D (CFD) ELISA Kit for detecting CFD in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.47-30ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P03953. Gene ID: 11537. Target Synonyms: CF-D; DF; PFD; ADN; Adipsin; Properdin Factor D; C3 convertase activator. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5923