Learn More
Biomatik Corporation Mouse Follicle Stimulating Hormone (FSH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04248
Encompass_Preferred
Mouse Follicle Stimulating Hormone (FSH) ELISA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.22-180ng/ml. Sensitivity: 0.77ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01216. Gene ID: 12640. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2532