Learn More
Biomatik Corporation Rat Cross Linked N-Telopeptide Of Type I Collagen (NTXI) ELISA Kit
Supplier:  Biomatik Corporation EKU03511
Encompass_Preferred
Rat Cross Linked N-Telopeptide Of Type I Collagen (NTXI) ELISA Kit for detecting NTXI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.89ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P02454; P02466. Gene ID: 29393; 84352. Target Synonyms: NTX-I; NTX1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5933