Learn More
Biomatik Corporation Rat Follicle Stimulating Hormone (FSH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04249
Encompass_Preferred
Rat Follicle Stimulating Hormone (FSH) ELISA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.94ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P11962; P18427. Gene ID: 116700; 25447. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2533