Learn More
Biomatik Corporation Mouse Cytochrome P450 3A4 (CYP3A4) ELISA Kit
Supplier:  Biomatik Corporation EKU03608
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 1.56-100ng/mL. Sensitivity: 0.57ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: CYP3A3; CYPIIIA4; 1,8-cineole 2-exo-monooxygenase; Albendazole sulfoxidase; Nifedipine oxidase; Quinine 3-monooxygenase; Taurochenodeoxycholate 6-alpha-hydroxylase. Restriction: For Research Use Only.
Catalog No. 50-228-5944