Learn More
Biomatik Corporation Rat Follistatin Like Protein 1 (FSTL1) ELISA Kit, 96 tests
Catalog No. 501502536
Encompass_Preferred
Rat Follistatin Like Protein 1 (FSTL1) ELISA Kit for detecting FSTL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62632. Gene ID: 79210. Target Synonyms: FRP; FSL1; Follistatin-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2536 Supplier Biomatik Corporation Supplier No. EKU04256
Rat Follistatin Like Protein 1 (FSTL1) ELISA Kit for detecting FSTL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62632. Gene ID: 79210. Target Synonyms: FRP; FSL1; Follistatin-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.