Learn More
Biomatik Corporation Mouse Follistatin Like Protein 3 (FSTL3) ELISA Kit, 96 tests
Catalog No. 501502537
Encompass_Preferred
Mouse Follistatin Like Protein 3 (FSTL3) ELISA Kit for detecting FSTL3 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EQC7. Gene ID: 83554. Target Synonyms: FLRG; FSRP; Follistatin-Related Protein,Secreted Glycoprotein; Follistatin-related gene protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2537 Supplier Biomatik Corporation Supplier No. EKU04257
Mouse Follistatin Like Protein 3 (FSTL3) ELISA Kit for detecting FSTL3 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EQC7. Gene ID: 83554. Target Synonyms: FLRG; FSRP; Follistatin-Related Protein,Secreted Glycoprotein; Follistatin-related gene protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.