Learn More
Biomatik Corporation Human Forkhead Box Protein O1 (FOXO1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04259
Encompass_Preferred
The Human Forkhead Box Protein O1 (FOXO1) ELISA Kit allows for quantitative determination of Forkhead Box Protein O1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.128 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q12778. Cross Activity: No significant cross-reactivity or interference between Forkhead Box Protein O1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2538