Learn More
Biomatik Corporation Mouse Forkhead Box Protein P3 (FOXP3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04263
Encompass_Preferred
Mouse Forkhead Box P3 (FOXP3) ELISA Kit for detecting FOXP3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99JB6. Gene ID: 20371. Target Synonyms: FOX-P3; AIID; DIETER; IPEX; JM2; PIDX; XPID; SCURFIN; Forkhead Box Protein P3; Immune Dysregulation,Polyendocrinopathy,Enteropathy,X-Linked. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2539