Learn More
Biomatik Corporation Mouse Elastase 2A (ELA2A) ELISA Kit
Supplier:  Biomatik Corporation EKU03844
Encompass_Preferred
Mouse Elastase 2A (ELA2A) ELISA Kit for detecting ELA2A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 14.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05208. Gene ID: 13706. Target Synonyms: ELA1; PE-1; CELA2A; Chymotrypsin-Like Elastase Family Member 2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5973