Learn More
Biomatik Corporation Mouse Elastin (ELN) ELISA Kit
Supplier:  Biomatik Corporation EKU03853
Encompass_Preferred
Mouse Elastin (ELN) ELISA Kit for detecting ELN in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P54320. Gene ID: 13717. Target Synonyms: WBS, WS, SVAS; Tropoelastin; Supravalvular Aortic Stenosis; Williams-Beuren Syndrome. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5975