Learn More
Biomatik Corporation Mouse Frataxin (FXN) ELISA Kit, 96 tests
Catalog No. 501502541
Encompass_Preferred
Mouse Frataxin (FXN) ELISA Kit for detecting FXN in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35943. Gene ID: 14297. Target Synonyms: FA; CyaY; FARR; FRDA; X25; Friedreich ataxia protein; Frataxin intermediate form; Frataxin, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2541 Supplier Biomatik Corporation Supplier No. EKU04270
Mouse Frataxin (FXN) ELISA Kit for detecting FXN in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35943. Gene ID: 14297. Target Synonyms: FA; CyaY; FARR; FRDA; X25; Friedreich ataxia protein; Frataxin intermediate form; Frataxin, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.