Learn More
Biomatik Corporation Human Elastin Microfibril Interface Located Protein 1 (EMILIN1) ELISA Kit
Supplier:  Biomatik Corporation EKU03855
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 1.56-100ng/mL. Sensitivity: 0.55ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9Y6C2. Target Synonyms: EMI; gp115; Elastin Microfibril Interfacer 1. Restriction: For Research Use Only.
Catalog No. 50-228-5977