Learn More
Biomatik Corporation Mouse Free Fatty Acid Receptor 3 (FFAR3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04273
Encompass_Preferred
Mouse Free Fatty Acid Receptor 3 (FFAR3) ELISA Kit for detecting FFAR3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3UFD7. Gene ID: 233080. Target Synonyms: FFA3R; GPR41; G Protein-Coupled Receptor 41. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2542