Learn More
Biomatik Corporation Mouse Endostatin (ES) ELISA Kit
Supplier:  Biomatik Corporation EKU03879
Encompass_Preferred
Mouse Endostatin (ES) ELISA Kit for detecting ES in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P39061. Gene ID: 12822. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5979